Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myeloid Specific Antigen Monoclonal Antibody (OTI4H6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526846
Description
Myeloid Specific Antigen Monoclonal Antibody for Western Blot, IHC (P)
This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
Specifications
Myeloid Specific Antigen | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P41218 | |
MNDA | |
Human recombinant protein fragment corresponding to amino acids 37-321 of MNDA produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4H6 | |
Unconjugated | |
MNDA | |
Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4332 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction