Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myeloid Specific Antigen Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556965 Supplier Invitrogen™ Supplier No. PA556965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQTSSSTPSN TSFTPNQETQ AQRQVDARRN VPQNDPVTVV VLKATAPFKY ESPENGKSTM FHATVASKTQ YFHVKVFDIN LKEKFVRKKV ITISDYSECK GVMEIKEASS VSDFN Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 50%.

This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloid Specific Antigen
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MNDA
Gene Accession No. P41218
Gene Alias Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135
Gene Symbols MNDA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Myeloid Specific Antigen. Recombinant protein control fragment (Product #RP-102257).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.