Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myeloperoxidase Monoclonal Antibody (8F4), Biotin
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549073
Description
Myeloperoxidase Monoclonal Antibody for ICC/IF, IHC (F), Flow, IA
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
Myeloperoxidase | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P11247 | |
MPO | |
Purified mouse MPO from WEHI-3 cell. | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Immunocytochemistry | |
8F4 | |
Biotin | |
MPO | |
84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
Mouse | |
Protein G | |
RUO | |
17523 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction