Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myeloperoxidase Monoclonal Antibody (9B12G7,4D8B12,9B12D9,9C11A5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515480
Description
MA5-15480 targets Myeloperoxidase in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15480 immunogen is purified recombinant fragment of MPO (aa1-193) expressed in E. Coli.
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
Myeloperoxidase | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P05164 | |
MPO | |
Purified recombinant fragment of MPO (aa1-193) expressed in E. Coli. | |
RUO | |
4353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
9B12G7,4D8B12,9B12D9,9C11A5, 4D8B12, 9B12D9, 9C11A5 | |
Unconjugated | |
MPO | |
84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction