Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase/MPO Antibody (2C7), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064803AF594
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Myeloperoxidase/MPO | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MPO | |
Human Myeloperoxidase | |
0.1 mL | |
Primary | |
NB100-64803 recognizes human myeloperoxidase (MPO). MPO is an important component of azurophilic granules in neutrophils, being involved in microbicidal processes. The protein is a multimer of 2 heavy chains (55kD) and two light chains (15kD), the heavy chains being linked by a disulphide bond. NB100-64803 recognizes native MPO in Western blots, and the heavy chain following boiling of the sample. The antibody also recognizes recombinant MPO in Western blots and weakly in ELISA. The antibody may be of value in the study of myeloid cells and myeloid leukaemias by flow cytometry following cell permeablisation. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
2C7 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
Mouse | |
Protein G purified | |
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
4353.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction