Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase/MPO Antibody (8F4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NBP151148
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.Specifications
Myeloperoxidase/MPO | |
Monoclonal | |
0.1 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
MPO | |
Purified mouse MPO from WEHI-3 cells | |
0.025 mg | |
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
4353 | |
Store at 4°C. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
8F4 | |
Unconjugated | |
EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction