Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase/MPO Antibody (MPO421-8B2), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241406V
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Flow Cytometry, ELISA, Flow (Intracellular).Specifications
Myeloperoxidase/MPO | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MPO | |
Human myeloperoxidase | |
0.1 mL | |
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
4353 | |
Human | |
Purified |
Flow Cytometry, ELISA | |
MPO421-8B2 | |
Flow Cytometry, ELISA, Flow (Intracellular) | |
EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody MPO421-8B2 recognizes human myeloperoxidase, a heme protein present in myeloblasts, neutrophils and monocytes. It is a marker of acute myelogenous leukemias and acute lymphoblastic leukemias. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction