Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase/MPO Antibody (RM407), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289098
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Myeloperoxidase/MPO | |
Monoclonal | |
Unconjugated | |
PBS, 50% Glycerol, 1% BSA | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM407 | |
Western Blot 1:200 - 1:500, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
A peptide corresponding to the internal region of light chain on MPO. This antibody may also react to mouse or rat MPO, as predicted by immunogen homology. | |
100 μL | |
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
4353 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction