Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase/MPO Antibody (rMPO/8694), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320650100UG
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Myeloperoxidase/MPO | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| Recombinant fragment (around aa150-250) of human Myeloperoxidase/MPO protein (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 4353 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rMPO/8694 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction