Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myeloperoxidase Recombinant Rabbit Monoclonal Antibody (6Z3W9)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542652
This item is not returnable.
View return policy
Description
Positive test controls include: HL-60. Immunogen sequence: GVSEPLKRKG RVGPLLACII GTQFRKLRDG DRFWWENEGV FSMQQRQALA QISLPRIICD NTGITTVSKN NIFMSNSYPR DFVNCSTLPA LNLASWREAS.
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
Myeloperoxidase | |
Recombinant Monoclonal | |
1.1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P05164 | |
MPO | |
A synthetic peptide corresponding to a sequence within amino acids 646-745 of human Myeloperoxidase (MPO) (NP_0002411). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
6Z3W9 | |
Unconjugated | |
MPO | |
84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4353 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction