Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYEOV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553016 Supplier Invitrogen™ Supplier No. PA553016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPALPIGLCT RCCLCLEQSP SWCHCLRGVS FLTFHLHQSV PLGDRDSLLM FTRQAGHFVE GSKAGRSRGR LCLSQALRVA VRGAFVSLWF AAGAGDRERN KGDKGAQTGA GLSQEAEDVD VSRARRVTDA PQGTLCGTGN Highest antigen sequence identity to the following orthologs: Mouse - 19%, Rat - 21%.

MYEOV, also named as OCIM, was originally isolated by the application of the NIH/3T3 tumorigenicity assay with DNA from a gastric carcinoma. It is a prognostic factor for patients with multiple myeloma, in part through a role of MYEOV in the control of MMC proliferation. MYEOV is a candidate oncogene activated in the amplification core located proximal toCCND1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYEOV
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYEOV
Gene Accession No. Q96EZ4
Gene Alias myeloma overexpressed; myeloma overexpressed (in a subset of t(11;14) positive multiple myelomas); myeloma overexpressed gene (in a subset of t(11;14) positive multiple myelomas); myeloma-overexpressed gene protein; MYEOV; OCIM; Oncogene in multiple myeloma
Gene Symbols MYEOV
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYEOV. Recombinant protein control fragment (Product #RP-89878).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.