Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYF-5 Antibody (593128), FITC, Novus Biologicals™
SDP

Catalog No. NB371008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371008 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371008 Supplier Novus Biologicals Supplier No. FAB4027F
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYF-5 Monoclonal antibody specifically detects MYF-5 in Human, Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen MYF-5
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 593128
Conjugate FITC
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, myogenic factor 5
Host Species Mouse
Immunogen E. coli-derived recombinant human MYF-5, Met1-Leu255, Accession # P13349
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4617
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.