Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYF-5 Antibody (593128), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4027P
Description
MYF-5 Monoclonal antibody specifically detects MYF-5 in Human, Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
MYF-5 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4617 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
593128 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, myogenic factor 5 | |
E. coli-derived recombinant human MYF-5, Met1-Leu255, Accession # P13349 | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction