Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYF-5 Antibody (OTI2G5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272855AF647
Description
MYF-5 Monoclonal specifically detects MYF-5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MYF-5 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYF5 | |
Full length recombinant protein of human MYF-5 (NP_005584) produced in E.coli. | |
0.1 mL | |
4617.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2G5 | |
Western Blot, Immunohistochemistry | |
BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, myogenic factor 5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction