Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYF-5 Antibody (SR1621), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32221850UL
Description
MYF-5 Monoclonal antibody specifically detects MYF-5 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
MYF-5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
4617 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
SR1621 | |
Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, myogenic factor 5 | |
A synthesized peptide derived from human MYF-5 (Uniprot #: P13349) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction