Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myf-5 Monoclonal Antibody (1E2G4), Active Motif™
SDP

Catalog No. 501992865
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2865 100 μg
50-199-2866 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2865 Supplier Active Motif Supplier No. 39801
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MYF5 is a member of the myogenic basic helix-loop-helix family of transcription factors, which can activate the muscle differentiation program.

Specifications

Antigen Myf-5
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1E2G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene MYF5
Gene Accession No. P24699
Gene Alias B130010J22Rik; basic helix-loop-helix nuclear protein; BHLHC2; BOS_4874; cb641; Class C basic helix-loop-helix protein 2; LOC100351906; muscle regulatory protein Myf5; Myf5; Myf-5; myf5 protein; MyoD family; Myogenic factor 5; myogenic factor 5-like; myogenic factor-5; myogenic regulatory factor 5; myogenic regulatory factor MYF-5; zgc:110509
Gene Symbols MYF5
Host Species Rat
Immunogen This Myf-5 antibody was raised against a peptide derived from mouse Myf-5.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17877
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.