Learn More
Invitrogen™ MYF5 Polyclonal Antibody

Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Rabbit (100%), Bovine (100%). Positive Control: human muscle. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MYF5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.03 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | MYF5 |
| Gene Accession No. | P13349 |
| Gene Alias | B130010J22Rik; basic helix-loop-helix nuclear protein; bHLHc2; cb641; Class C basic helix-loop-helix protein 2; muscle regulatory protein Myf5; MYF5; myf-5; myf5 protein; MyoD family; myogenic factor 5; myogenic factor-5; myogenic regulatory factor 5; myogenic regulatory factor MYF-5; unnamed protein product; zgc:110509 |
| Gene Symbols | MYF5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.