Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144924
Description
Immunogen sequence: FETGSYFFYL DGENVTLQPL EVAEGSPLYP GSDGTLSPCQ DQMPPEAGSD SSGEEHVLAP Antigen sequence identity: Mouse 98%; Rat 98%.
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
Specifications
MYF6 | |
Polyclonal | |
Unconjugated | |
Myf6 | |
bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4618 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23409 | |
Myf6 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 6-65. Recombinant protein control fragment (Product #RP-110125). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction