Learn More
Invitrogen™ MYF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120339
Description
Positive test controls include: HepG2, Mouse skeletal muscle, Mouse spleen, Rat skeletal muscle. The target is usually found in the following locations: Nucleus. Immunogen sequence: WACKTCKRKS APTDRRKAAT LRERRRLKKI NEAFEALKRR TVANPNQRLP KVEILRSAIS YIERLQDLLH RLDQQEKMQE LGVDPFSYRP KQENLEGADF L.
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
Specifications
| MYF6 | |
| Polyclonal | |
| Unconjugated | |
| Myf6 | |
| bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17878, 25714, 4618 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.02 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P15375, P19335, P23409 | |
| Myf6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-180 of human MYF6 (NP_002460.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.