Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH-2 Rat anti-Rat, Mouse, Clone: 8F72C8, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT848100UG
Description
Anti-MYH-2, clone 8F72C8 is a rat monoclonal antibody that detects Myosin-2 and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Specifications
MYH-2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
MYH2,MYHSA2 | |
KLH-conjugated linear peptide corresponding to 29 amino acids from the C-terminal half of human Myosin-2 | |
100 μg | |
Cell Structure | |
NP_001093582 | |
Rat, Mouse | |
Purified |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
8F72C8 | |
Unconjugated | |
Rat | |
Protein G Purified | |
RUO | |
Primary | |
Clone 8F72C8 is a rat monoclonal antibody that detects Myosin-2 in mouse and rat muscle. It targets an epitope within 29 amino acids from the C-terminal half. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction