Learn More
Invitrogen™ MYH10 Monoclonal Antibody (GT324)

Description
Positive Control: HeLa, MYH10-transfected 293T, NIH-3T3, mouse brain Predicted Reactivity: Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MYH10 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | GT324 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | MYH10 |
| Gene Accession No. | P35580, Q61879, Q9JLT0 |
| Gene Alias | 5730504C04Rik; 9330167F11Rik; cellular myosin heavy chain, type B; flectin; Fltn; MCH-B; MCH-B(B2); mKIAA3005; MYH10; Myhn2; Myhn-2; Myosin heavy chain 10; myosin heavy chain 10, non-muscle; myosin heavy chain, non-muscle IIb; myosin heavy chain, nonmuscle type B; myosin heavy chain, nonmuscular, type B; myosin IIB; myosin, heavy chain 10, non-muscle; myosin, heavy polypeptide 10, non-muscle; myosin-10; NMHC II-B; NMHC-B; NMHCII-B; NMMHC IIB; NMMHC II-b; NMMHCB; NMMHC-B; NMMHC-IIB; nonmuscle myosin; Non-muscle myosin heavy chain B; nonmuscle myosin heavy chain IIB; non-muscle myosin heavy chain IIb; nonmuscle myosin heavy chain II-B; nonmuscle myosin heavy chain-B; nonmuscle myosin II heavy chain-B; OTTHUMP00000165799; SMemb |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.