Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYH10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587092
Description
Purity is > 95% by SDS-PAGE.
MYH10 encodes a member of the myosin superfamily. The protein represents a conventional non-muscle myosin; it should not be confused with the unconventional myosin-10 (MYO10). Myosins are actin-dependent motor proteins with diverse functions including regulation of cytokinesis, cell motility, and cell polarity. Mutations in this gene have been associated with May-Hegglin anomaly and developmental defects in brain and heart. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
MYH10 | |
Polyclonal | |
Unconjugated | |
MYH10 | |
5730504C04Rik; 9330167F11Rik; cellular myosin heavy chain, type B; flectin; Fltn; MCH-B; MCH-B(B2); mKIAA3005; MYH10; Myhn2; Myhn-2; Myosin heavy chain 10; myosin heavy chain 10, non-muscle; myosin heavy chain, non-muscle IIb; myosin heavy chain, nonmuscle type B; myosin heavy chain, nonmuscular, type B; myosin IIB; myosin, heavy chain 10, non-muscle; myosin, heavy polypeptide 10, non-muscle; myosin-10; NMHC II-B; NMHC-B; NMHCII-B; NMMHC IIB; NMMHC II-b; NMMHCB; NMMHC-B; NMMHC-IIB; nonmuscle myosin; Non-muscle myosin heavy chain B; nonmuscle myosin heavy chain IIB; non-muscle myosin heavy chain IIb; nonmuscle myosin heavy chain II-B; nonmuscle myosin heavy chain-B; nonmuscle myosin II heavy chain-B; OTTHUMP00000165799; SMemb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4628, 77579, 79433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P35580, Q61879, Q9JLT0 | |
MYH10 | |
Recombinant fusion protein of human MYH10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction