Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116524 Supplier Invitrogen™ Supplier No. PA5116524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LO2, A375, Mouse liver, Mouse brain, Rat liver Immunogen sequence: ATAKENEKKA KSLEADLMQL QEDLAAAERA RKQADLEKEE LAEELASSLS GRNALQDEKR RLEARIAQLE EELEEEQGNM EAMSDRVRKA TQQAEQLSNE LATERSTAQK NESARQQLER QNKELRSKLH EMEGAVKSKF KSTIAALEAK IAQLEEQVEQ EAREKQAATK SLKQKDKKLK EILLQVEDER KMAEQYKEQA EKGNARVKQL KRQLEEAEEE SQRINANRRK LQRELDEATE SNEAMGREVN ALKSKLRRGN ETSFVPSRRS GGRRVIENAD GSEEETDTRD ADFNGTKASE.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH11
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MYH11
Gene Accession No. O08638, P35749, Q63862
Gene Alias AAT4; AV071570; contractile protein; FAA4; KIAA0866; LOW QUALITY PROTEIN: myosin-11; Myh11; myh11 {ECO:0000312; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; myosin-11; RGD:3136}; SM1; SM2; SMHC; SMMHC; smooth muscle myosin heavy chain
Gene Symbols MYH11
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17880, 24582, 4629
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.