Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589942 Supplier Invitrogen™ Supplier No. PA589942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALSKSKSNI ERTCRTVEDQ FSEIKAKDEQ QTQLIHDLNM QKARLQTQNG ELSHRVEEKE SLISQLTKSK QALTQQLEEL K; Positive Samples: Mouse eye, Rat eye; Cellular Location: Cytoplasm, myofibril.

MYH13 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH13
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Myh13
Gene Alias EO Myosin; extraocular muscle myosin heavy chain; extraocular myosin; extraocular myosin heavy chain; fast myosin heavy chain extraocular; MHCEO; MYH13; MyHC; MyHC-EO; MyHC-IIL; Myosin heavy chain 13; Myosin heavy chain, skeletal muscle, extraocular; Myosin heavy chain, skeletal muscle, laryngeal; myosin heavy polypeptide 13; myosin, heavy chain 13, skeletal muscle; myosin, heavy polypeptide 13, skeletal muscle; myosin-13; myosin-13; LOW QUALITY PROTEIN: myosin-13; skeletal muscle myosin heavy chain MyHC-EO/IIL; superfast myosin
Gene Symbols Myh13
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29605, 544791
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.