Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143724 Supplier Invitrogen™ Supplier No. PA5143724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 215 kDa* protein corresponding to the apparent molecular mass of MYH13 on SDS-PAGE immunoblots of mouse extraocular muscle, and does not detect MYHs in mouse diaphragm, gastrocnemius, or brain.

MYH13 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH13
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Myh13
Gene Accession No. Q9UKX3
Gene Alias EO Myosin; extraocular muscle myosin heavy chain; extraocular myosin; extraocular myosin heavy chain; fast myosin heavy chain extraocular; MHCEO; MYH13; MyHC; MyHC-EO; MyHC-IIL; Myosin heavy chain 13; Myosin heavy chain, skeletal muscle, extraocular; Myosin heavy chain, skeletal muscle, laryngeal; myosin heavy polypeptide 13; myosin, heavy chain 13, skeletal muscle; myosin, heavy polypeptide 13, skeletal muscle; myosin-13; myosin-13; LOW QUALITY PROTEIN: myosin-13; skeletal muscle myosin heavy chain MyHC-EO/IIL; superfast myosin
Gene Symbols Myh13
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the hinge region from mouse MYH13/extraocular myosin. This sequence has 100% identity with rat and human MYH13, and has low homology to other myosin family members.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29605, 544791, 8735
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.