Learn More
Invitrogen™ MYH13 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | MYH13 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Myh13 |
| Gene Accession No. | Q9UKX3 |
| Gene Alias | EO Myosin; extraocular muscle myosin heavy chain; extraocular myosin; extraocular myosin heavy chain; fast myosin heavy chain extraocular; MHCEO; MYH13; MyHC; MyHC-EO; MyHC-IIL; Myosin heavy chain 13; Myosin heavy chain, skeletal muscle, extraocular; Myosin heavy chain, skeletal muscle, laryngeal; myosin heavy polypeptide 13; myosin, heavy chain 13, skeletal muscle; myosin, heavy polypeptide 13, skeletal muscle; myosin-13; myosin-13; LOW QUALITY PROTEIN: myosin-13; skeletal muscle myosin heavy chain MyHC-EO/IIL; superfast myosin |
| Gene Symbols | Myh13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.