Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH14 Mouse anti-Human, Mouse, Clone: 1B4C9, Proteintech
SDP

Catalog No. 501737215 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7215 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7215 Supplier Proteintech Group Inc Supplier No. 668251IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.

Specifications

Antigen MYH14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B4C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MYH14
Gene Accession No. Q6URW6, Q7Z406
Gene Alias DFNA4, KIAA2034, MHC16, MYH14, myosin, Myosin 14, Myosin heavy chain 14, myosin, heavy chain 14, NMHC II C
Gene Symbols Myh14
Host Species Mouse
Immunogen MYH14 Fusion Protein Ag28063
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71960, 79784
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.