Learn More
MYH14/Myosin-14 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305270AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.Specifications
MYH14/Myosin-14 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q7Z406 | |
MYH14 | |
Between 1850 and 1900 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MYH14 | |
Myosin-14, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, Non-muscle myosin heavy chain IIc, NMHC II-C, MYH14, KIAA2034 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79784 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.