Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH14 Recombinant Rabbit Monoclonal Antibody (10E9C6)
GREENER_CHOICE

Catalog No. FEMA551391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551391 Supplier Invitrogen™ Supplier No. MA551391
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: AGKVDYKANE WLMKNMDPLN DNVAALLHQS TDRLTAEIWK DVEGIVGLEQ VSSLGDGPPG GRPRRGMFRT VGQLYKESLS RLMATLSNTN PSFVRCIVPN H.

Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH14
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10E9C6
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MYH14
Gene Accession No. Q6URW6, Q7Z406
Gene Alias 2400004E04Rik; DFNA4; DFNA4A; FP17425; II-C; KIAA2034; MHC16; Myh14; MYH14 variant protein; MYH17; myosin; myosin heavy chain 14; Myosin heavy chain, non-muscle IIc; myosin, heavy chain 14; myosin, heavy chain 14, non-muscle; myosin, heavy polypeptide 14; myosin-14; NHMCII; NMHC II-C; NMHC-II-C; nonmuscle myosin heavy chain; Non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; PNMHH
Gene Symbols MYH14
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 600-700 of human MYH14 (NP_079005.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308572, 71960, 79784
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.