Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH2 Mouse anti-Human, Clone: 1F1B6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MYH2 |
---|---|
Clone | 1F1B6 |
Concentration | 0.9 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6645
![]() ![]() |
Proteintech Group Inc
662121IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to MYH2 and will not bind other myosins.
Muscle contraction. Required for cytoskeleton organization.Specifications
MYH2 | |
0.9 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MYH2, MYH2A, MYHas8, MyHC 2A, MyHC IIa, MYHSA2, Myosin 2, Myosin 2a, Myosin heavy chain 2, Myosin heavy chain 2a, Myosin heavy chain IIa | |
MYH2 | |
IgG2b | |
Protein A | |
Antibody |
1F1B6 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UKX2 | |
4620 | |
Peptide | |
Primary | |
-20°C | |
MYH2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title