Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH2 Polyclonal Antibody

Catalog No. PIPA5121752
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121752 20 μL
PIPA5119293 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121752 Supplier Invitrogen™ Supplier No. PA5121752
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYH2 Polyclonal Antibody for Western Blot, IHC (P)

MYH2 (Myosin-2) is associated with muscle contraction and is required for cytoskeleton organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MYH2
Gene Accession No. Q9TV63
Gene Alias 2A myosin heavy chain; fast 2a myosin heavy chain; IBM3; inclusion body myopathy 3, autosomal dominant; MHC2A; Myh1; MYH2; MYH2A; MYHas8; MyHC-2A; MyHC-IIa; MYHSA2; Myhs-f; Myhsf1; Myhs-f1; myosin heavy chain 2; Myosin heavy chain 2a; myosin heavy chain IIa; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin-2; MYPOP; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain
Gene Symbols MYH2
Host Species Rabbit
Immunogen Recombinant protein Myosin Heavy Chain 2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17882, 397256, 691644
Target Species Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.