Learn More
Invitrogen™ MYH2 Polyclonal Antibody

Description
Predicted to react with Mouse and Rat samples.
Specifications
Specifications
| Antigen | MYH2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MYH2 |
| Gene Accession No. | Q9UKX2 |
| Gene Alias | 2A myosin heavy chain; fast 2a myosin heavy chain; IBM3; inclusion body myopathy 3, autosomal dominant; MHC2A; Myh1; MYH2; MYH2A; MYHas8; MyHC-2A; MyHC-IIa; MYHSA2; Myhs-f; Myhsf1; Myhs-f1; myosin heavy chain 2; Myosin heavy chain 2a; myosin heavy chain IIa; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin-2; MYPOP; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain |
| Gene Symbols | MYH2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.