Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120340 Supplier Invitrogen™ Supplier No. PA5120340
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse skeletal muscle, Rat skeletal muscle. The target is usually found in the following locations: Cytoplasm, myofibril.

MYH2 (Myosin-2) is associated with muscle contraction and is required for cytoskeleton organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MYH2
Gene Accession No. Q9UKX2
Gene Alias 2A myosin heavy chain; fast 2a myosin heavy chain; IBM3; inclusion body myopathy 3, autosomal dominant; MHC2A; Myh1; MYH2; MYH2A; MYHas8; MyHC-2A; MyHC-IIa; MYHSA2; Myhs-f; Myhsf1; Myhs-f1; myosin heavy chain 2; Myosin heavy chain 2a; myosin heavy chain IIa; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin-2; MYPOP; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain
Gene Symbols MYH2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17882, 4620, 691644
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.