Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | MYH4 |
|---|---|
| Concentration | 0.36 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2726
![]()
|
Proteintech Group Inc
201401AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to MYH4.
Muscle contraction. Weiss A., J. Mol. Biol. 290:61-75(1999). Denis N. J., Proteomics 7:868-874(2007).Specifications
| MYH4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q5SX39, Q9Y623 | |
| 17884, 4622 | |
| Peptide | |
| Primary | |
| -20°C | |
| MYH4 |
| 0.36 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MYH2B, MYH4, MyHC 2B, MyHC IIb, Myosin 4, Myosin heavy chain 2b, Myosin heavy chain 4, Myosin heavy chain IIb | |
| Myh4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title