Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH6/7 Rabbit anti-Mouse, Rat, Clone: 6V0G1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | MYH6/7 |
---|---|
Clone | 6V0G1 |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
MYH6/7 Monoclonal antibody specifically detects MYH6/7 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MYH6/7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
ASD3, CMD1EE, CMD1S, CMH1, CMH14, MPD1, MYHC, myHC-alpha, MyHC-beta, myhc-slow, Myosin Heavy Chain 6, Myosin heavy chain 7, Myosin heavy chain slow isoform, Myosin Heavy Chain, Cardiac Muscle Alpha Isoform, Myosin heavy chain, cardiac muscle beta isoform, myosin-6, myosin-7, SPMD, SSS3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1400 of human MYH6/7 (P12883). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
6V0G1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4624 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title