Learn More
Invitrogen™ MYH6 Monoclonal Antibody (CL2155)

Description
Immunogen sequence: QVEEDKVNSL SKSKVKLEQQ VDDLEGSLEQ EKKVRMDLER AKRKLEGDLK LTQESIMDLE NDKLQLEEKL KKKEFDINQQ NSKIEDEQVL ALQLQKKLKE NQARIEELEE ELEAERTARA KVEKLRSDLS RELEEISERL EEA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%. Binds to an epitope located within the peptide sequence KLEQQVDDLEGSLEQ as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | MYH6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL2155 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MYH6 |
| Gene Accession No. | P13533 |
| Gene Alias | A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMH14; MYH6; MYHC; Myhca; Myhc-a; MyHC-alpha; myosin 6; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-6; SSS3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.