Learn More
Invitrogen™ MYH6 Monoclonal Antibody (GT5612)

Description
Predicted Reactivity: Human (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MYH6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | GT5612 |
| Concentration | 1.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | MYH6 |
| Gene Accession No. | P13533, Q02566 |
| Gene Alias | A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMH14; MYH6; MYHC; Myhca; Myhc-a; MyHC-alpha; myosin 6; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-6; SSS3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.