Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH6 Monoclonal Antibody (GT565)
GREENER_CHOICE

Catalog No. PIMA527820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527820 Supplier Invitrogen™ Supplier No. MA527820
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: mouse heart Predicted Reactivity: Human (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cardiac muscle myosin is a hexamer consisting of two heavy chain subunits, two light chain subunits, and two regulatory subunits. This gene encodes the alpha heavy chain subunit of cardiac myosin. The gene is located approximately 4kb downstream of the gene encoding the beta heavy chain subunit of cardiac myosin. Mutations in this gene cause familial hypertrophic cardiomyopathy and atrial septal defect 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH6
Applications Western Blot
Classification Monoclonal
Clone GT565
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MYH6
Gene Accession No. P13533, Q02566
Gene Alias A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMH14; MYH6; MYHC; Myhca; Myhc-a; MyHC-alpha; myosin 6; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-6; SSS3
Gene Symbols MYH6
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human MYH6. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17888, 4624
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.