Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551564 Supplier Invitrogen™ Supplier No. PA551564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVEEDKVNSL SKSKVKLEQQ VDDLEGSLEQ EKKVRMDLER AKRKLEGDLK LTQESIMDLE NDKLQLEEKL KKKEFDINQQ NSKIEDEQVL ALQLQKKLKE NQARIEELEE ELEAERTARA KVEKLRSDLS RELEEISERL EEA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Cardiac muscle myosin is a hexamer consisting of two heavy chain subunits, two light chain subunits, and two regulatory subunits. This gene encodes the alpha heavy chain subunit of cardiac myosin. The gene is located approximately 4kb downstream of the gene encoding the beta heavy chain subunit of cardiac myosin. Mutations in this gene cause familial hypertrophic cardiomyopathy and atrial septal defect 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYH6
Gene Accession No. P13533
Gene Alias A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMH14; MYH6; MYHC; Myhca; Myhc-a; MyHC-alpha; myosin 6; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-6; SSS3
Gene Symbols MYH6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYBPC2. Recombinant protein control fragment (Product #RP-89003).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.