Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH7 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297775FR
Description
MYH7 Polyclonal specifically detects MYH7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MYH7 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
4625 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
cardiac muscle beta isoform, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM | |
Produced in rabbits immunized with E. coli-derived Human MYH7 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction