Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH7 Monoclonal Antibody (C1), Invitrogen™

Catalog No. PIMA543924
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543924 20 μL
PIMA543923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543924 Supplier Invitrogen™ Supplier No. MA543924
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscle myosin is a hexameric protein containing 2 heavy chain subunits, 2 alkali light chain subunits, and 2 regulatory light chain subunits. This gene encodes the beta (or slow) heavy chain subunit of cardiac myosin. It is expressed predominantly in normal human ventricle. It is also expressed in skeletal muscle tissues rich in slow-twitch type I muscle fibers. Changes in the relative abundance of this protein and the alpha (or fast) heavy subunit of cardiac myosin correlate with the contractile velocity of cardiac muscle. Its expression is also altered during thyroid hormone depletion and hemodynamic overloading. Mutations in this gene are associated with familial hypertrophic cardiomyopathy, myosin storage myopathy, dilated cardiomyopathy, and Laing early-onset distal myopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH7
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Myh7
Gene Accession No. P02564, P12883, P79293
Gene Alias beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin; beta-myosin heavy chain; beta-myosin-heavy-chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain beta isoform; CMD1S; CMH1; LOC100727138; mg:cb02g011; MPD1; Myh7; MYH-beta/slow; MyHC-alpha; Myhcb; Myhc-b; myHC-beta; MyHC-I; MyHC-slow; myopathy, distal 1; myosin beta heavy chain; myosin heavy chain 1; myosin heavy chain 7; myosin heavy chain cardiac alpha; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow; myosin heavy chain slow isoform; myosin heavy chain slow type 1 (beta cardiac); myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 7; myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy polypeptide 7, cardiac muscle, beta; myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A; slow myosin heavy chain; smyhc3; SPMD; SPMM; type slow/beta myosin heavy chain; Unknown (protein for IMAGE:7985957); ventricular myosin heavy chain; vmhc
Gene Symbols Myh7
Host Species Mouse
Immunogen Ala1777-Glu1935 (Accession P12883), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29557, 396860, 4625
Target Species Human, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.