Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100023 Supplier Invitrogen™ Supplier No. PA5100023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYH7.

Muscle myosin is a hexameric protein containing 2 heavy chain subunits, 2 alkali light chain subunits, and 2 regulatory light chain subunits. This gene encodes the beta (or slow) heavy chain subunit of cardiac myosin. It is expressed predominantly in normal human ventricle. It is also expressed in skeletal muscle tissues rich in slow-twitch type I muscle fibers. Changes in the relative abundance of this protein and the alpha (or fast) heavy subunit of cardiac myosin correlate with the contractile velocity of cardiac muscle. Its expression is also altered during thyroid hormone depletion and hemodynamic overloading. Mutations in this gene are associated with familial hypertrophic cardiomyopathy, myosin storage myopathy, dilated cardiomyopathy, and Laing early-onset distal myopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Myh7
Gene Accession No. P02564, P12883, Q91Z83
Gene Alias beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin; beta-myosin heavy chain; beta-myosin-heavy-chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain beta isoform; CMD1S; CMH1; LOC100727138; mg:cb02g011; MPD1; Myh7; MYH-beta/slow; MyHC-alpha; Myhcb; Myhc-b; myHC-beta; MyHC-I; MyHC-slow; myopathy, distal 1; myosin beta heavy chain; myosin heavy chain 1; myosin heavy chain 7; myosin heavy chain cardiac alpha; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow; myosin heavy chain slow isoform; myosin heavy chain slow type 1 (beta cardiac); myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 7; myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy polypeptide 7, cardiac muscle, beta; myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A; slow myosin heavy chain; smyhc3; SPMD; SPMM; type slow/beta myosin heavy chain; Unknown (protein for IMAGE:7985957); ventricular myosin heavy chain; vmhc
Gene Symbols Myh7
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYH7(Accession P12883), corresponding to amino acid residues E1705-A1755.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140781, 29557, 4625
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.