Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH7B Antibody (SR1337), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MYH7B |
---|---|
Clone | SR1337 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
MYH7B Monoclonal antibody specifically detects MYH7B in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MYH7B | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
57644 | |
IgG | |
Affinity purified |
SR1337 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Antigen MLAA-21, MHC14, MYH14, Myosin cardiac muscle beta chain, myosin heavy chain 7B, Myosin heavy chain 7B, cardiac muscle beta isoform, Myosin-7B, Slow A MYH14 | |
A synthesized peptide derived from human MYH7B (Uniprot #: A7E2Y1) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title