Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143722 Supplier Invitrogen™ Supplier No. PA5143722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 215 kDa* protein corresponding to the apparent molecular mass of MYH7B on SDS-PAGE immunoblots of mouse extraocular muscle and brain.

Myosin cardiac muscle beta chain or MYH7B is believed to be involved in cardiac slow-twitch myosin contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH7B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene MYH7B
Gene Accession No. A2AQP0, A7E2Y1
Gene Symbols MYH7B
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the hinge region from mouse MYH7B/MHC14. This sequence is highly conserved in rat and human MYH7B, and has less than 50% identity with other MYH family members.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311570, 57644, 668940
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.