Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH7B Recombinant Rabbit Monoclonal Antibody (7W8J3)
GREENER_CHOICE

Catalog No. PIMA542691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542691 Supplier Invitrogen™ Supplier No. MA542691
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse skeletal muscle, Rat skeletal muscle, Rat heart. Immunogen sequence: LAEQELLEAT ERLNLLHSQN TGLLNQKKKL EADLAQLSGE VEEAAQERRE AEEKAKKAIT DAAMMAEELK KEQDTSAHLE RMKKTLEQTV RELQARLEEA EQAALRGGKK QVQKLEAKVR ELEAELDAEQ KKHAEALKGV RKHERRVKEL AYQAEEDRKN LARMQDLVDK LQSKVKSYKR QFEEAEQQAN TNLAKYRKAQ HELDDAEERA DMAETQANKL RARTRDALGP KHKE.

Myosin cardiac muscle beta chain or MYH7B is believed to be involved in cardiac slow-twitch myosin contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH7B
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 7W8J3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYH7B
Gene Accession No. A2AQP0
Gene Symbols MYH7B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1750-1983 of human MYH7B (NP_0659353).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311570, 668940
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.