Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYH8 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP241309
Description
MYH8 Polyclonal specifically detects MYH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MYH8 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
MYH8 | |
Antibody was raised against a 19 amino acid peptide near the amino terminus of human MYH8. The immunogen is located within amino acids 60 - 110 of MYH8. Amino Acid Squence: VREDQVFPMNPPKYDKIED | |
Affinity Purified | |
RUO | |
4626 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL | |
Fetal-Myosin Heavy Chain, gtMHC-F, MyHC-peri, myHC-perinatal, MyHC-pn, Myosin Heavy Chain 8, Myosin Heavy Chain, Skeletal Muscle, Perinatal, Myosin, Heavy Chain 8, Skeletal Muscle, Perinatal, Myosin, Heavy Polypeptide 8, Skeletal Muscle, Perinatal, myosin-8 | |
Rabbit | |
220 kDa | |
0.1 mg | |
Primary | |
MYH8 antibody is predicted to not cross-react with other members of the myosin heavy chain family. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction