Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537180 Supplier Invitrogen™ Supplier No. PA537180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 223 kDa. Purity is >95% by SDS-PAGE.

Myosin is a highly conserved, ubiquitously expressed protein that interacts with Actin to generate the force for cellular movements. Conventional Myosins are hexameric proteins consisting of two heavy chain subunits, a pair of non-phosphorylatable light chain subunits and a pair of phosphorylatable light chain subunits. Three general classes of Myosin have been cloned: smooth muscle Myosins, striated muscle Myosins and non-muscle Myosins. Contractile activity in smooth muscle is regulated by the calcium/calmodulin-dependent phosphorylation of Myosin light chain (MLC) by Myosin light chain kinase. Myosin heavy chains, which are encoded by the MYH gene family, contain Actin-activated ATPase activity which generates the motor function of Myosin. Myosin heavy chains were initially isolated from a human fetal skeletal muscle and are the major determinant in the speed of contraction of skeletal muscle. Various isoforms of myosin heavy chains are differentially expressed depending on the functional activity of the muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MYH8
Gene Accession No. P04462, P13535, P13542
Gene Alias 4832426G23Rik; AI327267; Beta-MHC antisense; DA7; fetal-myosin heavy chain; gtMHC-F; MHCp; Myh8; MyHC-peri; MyHC-perinatal; MyHC-pn; Myhsp; Myhs-p; Myosin 8; Myosin heavy chain 8; myosin heavy chain, skeletal muscle, perinatal; myosin, heavy chain 8, skeletal muscle, perinatal; myosin, heavy polypeptide 8, skeletal muscle, perinatal; Myosin8; myosin-8
Gene Symbols MYH8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human Myosin-8.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17885, 252942, 4626
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.