Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH9 Antibody, ProSci
SDP

Catalog No. 16814611
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-814-611 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-814-611 Supplier ProSci LLC Supplier No. 130660100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYH9 Polyclonal Antibody specifically detects GDF15 in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MYH9
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution WB: 1:1000 - 1:2000, IHC: 1:50 - 1:100
Formulation PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
Gene Alias MYH9, DFNA17, EPSTS, FTNS, MGC104539, MHA, NMHC-II-A, NMMHCA, BDPLT6, NMMHC-IIA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1711-1960 of human MYH9 (NP_002464.1).
Purification Method Affinity purification
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4627
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze / thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Cell Cycle, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.