Learn More
MYL1 Mouse anti-Human, Clone: OTI1D9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810681

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.Specifications
MYL1 | |
Monoclonal | |
Unconjugated | |
MYL1 | |
MLC1F, MLC3F | |
Mouse | |
Affinity Chromatography | |
RUO | |
4632 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1D9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05976 | |
MYL1 | |
Human recombit protein fragment corresponding to amino acids 2-150 of human MYL1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.