Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597054 Supplier Invitrogen™ Supplier No. PA597054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSFSADQIAE FKEAFLLFDR TGDSKITLSQ VGDVLRALGT NPTNAEVRKV LGNPSNEELN AKKIEFEQFL PMMQAISNNK DQATYEDFVE GLRVFDKEGN GTVMGAELRH VLATLGEKMK EEEVEALMAG QEDSNGCINY EAFVKHIMSI; Positive Samples: HL-60, Mouse heart, Rat heart.

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYL1
Gene Accession No. P02600, P05976, P05977
Gene Alias A1 catalytic; A2 catalytic; AI325107; alkali myosin light chain 1; alkali myosin light chain 3; alkali myosin light chain 3 A2 catalytic; Ca2+-binding protein-like protein; fast myosin alkali light chain 1; fast myosin alkali light chain 3; fast myosin light chain 1; fast myosin light chain 1F; Mcl3; MLC; Mlc1; MLC1/MLC3; MLC1F; MLC1F/MLC3F; Mlc3; MLC3F; MLC3-f; MLCf; Myl1; Mylf; myosin light chain; myosin light chain 1; myosin light chain 1, skeletal muscle isoform; myosin light chain 1/3, skeletal muscle isoform; myosin light chain 3; myosin light chain 3, skeletal muscle isoform; myosin light chain A1/A2; Myosin light chain alkali 1/2; myosin light chain, alkali, fast skeletal muscle; myosin, light chain 1; myosin, light chain 1, alkali; skeletal, fast; myosin, light polypeptide 1; myosin, light polypeptide 1, alkali; atrial, embryonic; myosin, light polypeptide 1, alkali; skeletal, fast; myosin, light polypeptide 4, alkali; atrial, embryonic
Gene Symbols MYL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human MYL1 (NP_5241461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17901, 4632, 56781
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.